Blood collecting system, S-Monovette® metal analysis
Toimittaja: SARSTEDT
01.1604.400 85.1162.600
SARS01.1604.400EA 79.8 EUR
SARS01.1604.400 SARS85.1162.600
Blood collecting system, S-Monovette® metal analysis
Diagnostiset testit Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
S-Monovette® is a blood collection system that combines two blood collection techniques – aspiration and vacuum. The S-Monovette® is suitable for all vein conditions and achieves an optimal sample quality, thereby producing the best results. The aspiration technique is a gentle technique for routine blood collection. Using the vacuum technique, a 'fresh' vacuum is always available.
The S-Monovette® metal analysis, used in combination with a special S-Monovette® needle, was developed for testing the most frequently occurring trace elements. As an anticoagulant, lithium heparin (19 I.E./mlMore Product Information