Sartocon® Slice 200 Cassettes
Toimittaja: Sartorius
3081460902E-SG 3081465902E-SG 3081445902E-SW 3081443902E-SW 3081463902E-SW 3081465902E-SW
SARP3081460902E-SGEA 597 EUR
SARP3081460902E-SG SARP3081465902E-SG SARP3081445902E-SW SARP3081443902E-SW SARP3081463902E-SW SARP3081465902E-SW
Sartocon® Slice 200 Cassettes
Suodattimet Tangenttivirtauskasetit
Polyethersulfone membrane
The robust polyethersulfone membrane (PESU) is a polymer, stable within a broad pH (1 to 14) and temperature range. Membrane regeneration and depyrogenation is accomplished by using (1 N) NaOH at elevated temperatures. Cassettes are stored in 0,1 N NaOH.
  • Low hold-up volume and low protein-binding
  • Wide pH and temperature range
  • High flow rates
  • Silicone sealing compound
Hydrosart® membrane
The cellulose-based membrane is stable polymer, which is compatible with a wide range of chemical agents and stable within a broad pH range. ItMore Product Information