Kalibrointipainot, luokka F1, Proof Line
Toimittaja: Sartorius Balances
YCW413-00 YCW513-02 YCW413-02 YCW023-02 YCW153-00 YCW153-02 YCW213-00 YCW213-02 YCW023-00 YCW013-00 YCW013-02 YCW223-00 YCW423-00 YCW553-00 YCW353-00 YCW313-00 YCW323-02 YCW723-02 YCW653-00 YCW653-02 YCW713-02 YCW223-02 YCW123-00 YCW053-00 YCW113-02 YCW123-02 YCW313-02 YCW353-02 YCW513-00 YCW523-00 YCW723-00 YCW113-00 YCW253-00 YCW053-02 YCW253-02 YCW613-02 YCW423-02 YCW523-02 YCW623-02 YCW453-02 YCW553-02 YCW613-00 YCW323-00 YCW623-00 YCW453-00 YCW713-00 YCW313-AC-02 YCW353-AC-02 YCW353-AC-00 YCW313-AC-00 YCW323-AC-02 YCW323-AC-00 YCW623-AC-02 YCW623-AC-00 YCW653-AC-00 YCW613-AC-02 YCW613-AC-00 YCW713-AC-02 YCW713-AC-00 YCW653-AC-02 YCW553-AC-00 YCW423-AC-00 YCW453-AC-02 YCW453-AC-00 YCW423-AC-02 YCW413-AC-02 YCW413-AC-00 YCW523-AC-02 YCW553-AC-02 YCW513-AC-02 YCW513-AC-00 YCW523-AC-00
SARBYCW413-00EA 104 EUR
SARBYCW413-00 SARBYCW513-02 SARBYCW413-02 SARBYCW023-02 SARBYCW153-00 SARBYCW153-02 SARBYCW213-00 SARBYCW213-02 SARBYCW023-00 SARBYCW013-00 SARBYCW013-02 SARBYCW223-00 SARBYCW423-00 SARBYCW553-00 SARBYCW353-00 SARBYCW313-00 SARBYCW323-02 SARBYCW723-02 SARBYCW653-00 SARBYCW653-02 SARBYCW713-02 SARBYCW223-02 SARBYCW123-00 SARBYCW053-00 SARBYCW113-02 SARBYCW123-02 SARBYCW313-02 SARBYCW353-02 SARBYCW513-00 SARBYCW523-00 SARBYCW723-00 SARBYCW113-00 SARBYCW253-00 SARBYCW053-02 SARBYCW253-02 SARBYCW613-02 SARBYCW423-02 SARBYCW523-02 SARBYCW623-02 SARBYCW453-02 SARBYCW553-02 611-1707 SARBYCW323-00 611-1708 SARBYCW453-00 SARBYCW713-00 SARBYCW313-AC-02 SARBYCW353-AC-02 SARBYCW353-AC-00 SARBYCW313-AC-00 SARBYCW323-AC-02 SARBYCW323-AC-00 SARBYCW623-AC-02 SARBYCW623-AC-00 SARBYCW653-AC-00 SARBYCW613-AC-02 SARBYCW613-AC-00 SARBYCW713-AC-02 SARBYCW713-AC-00 SARBYCW653-AC-02 SARBYCW553-AC-00 SARBYCW423-AC-00 SARBYCW453-AC-02 SARBYCW453-AC-00 SARBYCW423-AC-02 SARBYCW413-AC-02 SARBYCW413-AC-00 SARBYCW523-AC-02 SARBYCW553-AC-02 SARBYCW513-AC-02 SARBYCW513-AC-00 SARBYCW523-AC-00
Kalibrointipainot, luokka F1, Proof Line
Painot Kalibrointipainot
Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.
  • 0 to 5 mg leaf weights: Aluminium
  • 10 to 500 mg leaf weights: Nickel silver
  • 1 g to 10 Kg knob weights: Stainless steel
Sertifikaatit: Compliance with OIML R111-1.