Calibration weight sets in wooden boxes, OIML class F2
Toimittaja: Sartorius Balances
YCS31-614-02 YCS31-654-00 YCS31-654-02 YCS01-654-02 YCS31-614-00 YCS01-654-00 YCS01-614-00 YCS31-714-00 YCS31-714-02 YCS01-524-00 YCS01-524-02 YCS01-514-00 YCS01-514-02 YCS01-614-02 YCS31-654-AC02 YCS01-254-AC02 YCS01-254-AC00 YCS31-654-AC00 YCS31-614-AC00 YCS31-514-AC02 YCS31-514-AC00 YCS31-614-AC02
SARBYCS31-614-02EA 2090 EUR
SARBYCS31-614-02 SARBYCS31-654-00 SARBYCS31-654-02 SARBYCS01-654-02 SARBYCS31-614-00 SARBYCS01-654-00 SARBYCS01-614-00 SARBYCS31-714-00 SARBYCS31-714-02 SARBYCS01-524-00 SARBYCS01-524-02 SARBYCS01-514-00 SARBYCS01-514-02 SARBYCS01-614-02 SARBYCS31-654-AC02 SARBYCS01-254-AC02 SARBYCS01-254-AC00 SARBYCS31-654-AC00 SARBYCS31-614-AC00 SARBYCS31-514-AC02 SARBYCS31-514-AC00 SARBYCS31-614-AC02
Calibration weight sets in wooden boxes, OIML class F2
Painot Kalibrointipainot
Class F2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.