Cubis® II MCE Ultra-High Resolution Semi-Micro Balances
Toimittaja: Sartorius Balances
MCE225S3CEI MCE125S3CEM MCE125S3CEI MCE225S3S0I MCE225S3S0M MCE125S3S0M MCE225P3CEI MCE225P3S0I MCE225P3S0M MCE225P3CEM MCE225S3CEM MCE125S3S0I
SARBMCE225S3CEIEA 23590 EUR
SARBMCE225S3CEI SARBMCE125S3CEM SARBMCE125S3CEI SARBMCE225S3S0I SARBMCE225S3S0M SARBMCE125S3S0M SARBMCE225P3CEI SARBMCE225P3S0I SARBMCE225P3S0M SARBMCE225P3CEM SARBMCE225S3CEM SARBMCE125S3S0I
Cubis® II MCE Ultra-High Resolution Semi-Micro Balances
Vaa'at
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCE display - StateMore Product Information