Analytical laboratory balances, Cubis® II MCE
Toimittaja: Sartorius Balances
MCE224S2S00-A MCE524S2S00-U MCE524P2S00-U MCE324S2S00-U MCE324P2S00-U MCE224S2S00-U MCE124S2S00-U MCE524S2S00-A MCE524P2S00-A MCE324S2S00-A MCE324P2S00-A MCE124S2S00-A MCE524S2S00-I MCE524P2S00-I MCE324S2S00-I MCE324P2S00-I MCE224S2S00-I MCE124S2S00-I MCE224S-U-MTS MCE225S-I-3MTS
SARBMCE224S2S00-AEA 13080 EUR
SARBMCE224S2S00-A SARBMCE524S2S00-U SARBMCE524P2S00-U SARBMCE324S2S00-U SARBMCE324P2S00-U SARBMCE224S2S00-U SARBMCE124S2S00-U SARBMCE524S2S00-A SARBMCE524P2S00-A SARBMCE324S2S00-A SARBMCE324P2S00-A SARBMCE124S2S00-A SARBMCE524S2S00-I SARBMCE524P2S00-I SARBMCE324S2S00-I SARBMCE324P2S00-I SARBMCE224S2S00-I SARBMCE124S2S00-I 611-5826 611-5832
Analytical laboratory balances, Cubis® II MCE
Vaa'at
Cubis® II MCE analytical balances with essential user interface offer a maximum weighing capacity between 120 and 520 g in combination with readability of 0.1 mg.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required toMore Product Information