Precision (milligram) Laboratory Balances, Cubis® II MCA
Toimittaja: Sartorius Balances
MCA623S2S00A99 MCA623P2S00A99 MCA323S2S00A99 MCA623S2S00E99 MCA623P2S00E99 MCA323S2S00E99 MCA623S2S00I99 MCA623P2S00I99 MCA323S2S00I99 MCA623S2S00R99 MCA623P2S00R99 MCA323S2S00R99 MCA623S2S00U99 MCA623P2S00U99 MCA323S2S00U99 MCA5203S2S00-E MCA5203P2S00-E MCA3203S2S00-E MCA2203S2S00-E MCA2203P2S00-E MCA1203S2S00-E MCA623S2S00-E MCA623P2S00-E MCA323S2S00-E MCA5203S2S00-R MCA5203P2S00-R MCA3203S2S00-R MCA2203S2S00-R MCA2203P2S00-R MCA1203S2S00-R MCA623S2S00-R MCA623P2S00-R MCA323S2S00-R MCA5202S2S00-E MCA5202S2S00-R MCA14202S2S000 MCA14202P2S000 MCA10202S2S000 MCA12201S2S000 MCA5203S2SA99 MCA5203P2SA99 MCA3203S2SA99 MCA2203S2SA99 MCA2203P2SA99 MCA1203S2SA99 MCA11201S2S000 MCA5203S2SE99 MCA5203P2SE99 MCA3203S2SE99 MCA2203S2SE99 MCA2203P2SE99 MCA1203S2SE99 MCA5203S2SI99 MCA5203P2SI99 MCA3203S2SI99 MCA2203S2SI99 MCA2203P2SI99 MCA1203S2SI99 MCA5203S2SR99 MCA5203P2SR99 MCA3203S2SR99 MCA2203S2SR99 MCA2203P2SR99 MCA1203S2SR99 MCA5203S2SU99 MCA5203P2SU99 MCA3203S2SU99 MCA2203S2SU99 MCA2203P2SU99 MCA1203S2SU99 MCA14202S2099 MCA14202P2099 MCA10202S2099 MCA8202S2S099 MCA6202S2S099 MCA6202P2S099 MCA4202S2S099 MCA2202S2S099 MCA1202S2S099 MCA12201S2099 MCA8201S2S099 MCA5201S2S099 MCA11201S2099 MCA5202S2SA99 MCA5202S2SE99 MCA5202S2SI99 MCA5202S2SR99 MCA5202S2SU99 MCA5203S2S00-A MCA5203P2S00-A MCA3203S2S00-A MCA2203S2S00-A MCA2203P2S00-A MCA1203S2S00-A MCA623S2S00-A MCA623P2S00-A MCA323S2S00-A MCA5203S2S00-I MCA5203P2S00-I MCA3203S2S00-I MCA2203S2S00-I MCA2203P2S00-I MCA1203S2S00-I MCA623S2S00-I MCA623P2S00-I MCA323S2S00-I MCA5203S2S00-U MCA5203P2S00-U MCA3203S2S00-U MCA2203S2S00-U MCA2203P2S00-U MCA1203S2S00-U MCA623S2S00-U MCA623P2S00-U MCA323S2S00-U MCA8202S2S00-0 MCA6202S2S00-0 MCA6202P2S00-0 MCA4202S2S00-0 MCA2202S2S00-0 MCA8201S2S00-0 MCA5202S2S00-A MCA5202S2S00-I MCA5202S2S00-U MCA5201S2S00-0 MCA1202S2CFR01 MCA5203S200RQP
SARBMCA623S2S00A99EA 15040 EUR
SARBMCA623S2S00A99 SARBMCA623P2S00A99 SARBMCA323S2S00A99 SARBMCA623S2S00E99 SARBMCA623P2S00E99 SARBMCA323S2S00E99 SARBMCA623S2S00I99 SARBMCA623P2S00I99 SARBMCA323S2S00I99 SARBMCA623S2S00R99 SARBMCA623P2S00R99 SARBMCA323S2S00R99 SARBMCA623S2S00U99 SARBMCA623P2S00U99 SARBMCA323S2S00U99 SARBMCA5203S2S00-E SARBMCA5203P2S00-E SARBMCA3203S2S00-E SARBMCA2203S2S00-E SARBMCA2203P2S00-E SARBMCA1203S2S00-E SARBMCA623S2S00-E SARBMCA623P2S00-E SARBMCA323S2S00-E SARBMCA5203S2S00-R SARBMCA5203P2S00-R SARBMCA3203S2S00-R SARBMCA2203S2S00-R SARBMCA2203P2S00-R SARBMCA1203S2S00-R SARBMCA623S2S00-R SARBMCA623P2S00-R SARBMCA323S2S00-R SARBMCA5202S2S00-E SARBMCA5202S2S00-R SARBMCA14202S2S000 SARBMCA14202P2S000 SARBMCA10202S2S000 SARBMCA12201S2S000 SARBMCA5203S2SA99 SARBMCA5203P2SA99 SARBMCA3203S2SA99 SARBMCA2203S2SA99 SARBMCA2203P2SA99 SARBMCA1203S2SA99 SARBMCA11201S2S000 SARBMCA5203S2SE99 SARBMCA5203P2SE99 SARBMCA3203S2SE99 SARBMCA2203S2SE99 SARBMCA2203P2SE99 SARBMCA1203S2SE99 SARBMCA5203S2SI99 SARBMCA5203P2SI99 SARBMCA3203S2SI99 SARBMCA2203S2SI99 SARBMCA2203P2SI99 SARBMCA1203S2SI99 SARBMCA5203S2SR99 SARBMCA5203P2SR99 SARBMCA3203S2SR99 SARBMCA2203S2SR99 SARBMCA2203P2SR99 SARBMCA1203S2SR99 SARBMCA5203S2SU99 SARBMCA5203P2SU99 SARBMCA3203S2SU99 SARBMCA2203S2SU99 SARBMCA2203P2SU99 SARBMCA1203S2SU99 SARBMCA14202S2099 SARBMCA14202P2099 SARBMCA10202S2099 SARBMCA8202S2S099 SARBMCA6202S2S099 SARBMCA6202P2S099 SARBMCA4202S2S099 SARBMCA2202S2S099 SARBMCA1202S2S099 SARBMCA12201S2099 SARBMCA8201S2S099 SARBMCA5201S2S099 SARBMCA11201S2099 SARBMCA5202S2SA99 SARBMCA5202S2SE99 SARBMCA5202S2SI99 SARBMCA5202S2SR99 SARBMCA5202S2SU99 SARBMCA5203S2S00-A SARBMCA5203P2S00-A SARBMCA3203S2S00-A SARBMCA2203S2S00-A SARBMCA2203P2S00-A SARBMCA1203S2S00-A SARBMCA623S2S00-A SARBMCA623P2S00-A SARBMCA323S2S00-A SARBMCA5203S2S00-I SARBMCA5203P2S00-I SARBMCA3203S2S00-I SARBMCA2203S2S00-I SARBMCA2203P2S00-I SARBMCA1203S2S00-I SARBMCA623S2S00-I SARBMCA623P2S00-I SARBMCA323S2S00-I SARBMCA5203S2S00-U SARBMCA5203P2S00-U SARBMCA3203S2S00-U SARBMCA2203S2S00-U SARBMCA2203P2S00-U SARBMCA1203S2S00-U SARBMCA623S2S00-U SARBMCA623P2S00-U SARBMCA323S2S00-U SARBMCA8202S2S00-0 SARBMCA6202S2S00-0 SARBMCA6202P2S00-0 SARBMCA4202S2S00-0 SARBMCA2202S2S00-0 SARBMCA8201S2S00-0 SARBMCA5202S2S00-A SARBMCA5202S2S00-I SARBMCA5202S2S00-U SARBMCA5201S2S00-0 611-5265 611-5266
Precision (milligram) Laboratory Balances, Cubis® II MCA
Vaa'at
Cubis® II MCA precision balances with advanced user interface offer a maximum load of between 320 g and 14200 g and a readability of 1 to 100 mg provide, ideal model for every application.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives andMore Product Information