High-capacity toploading laboratory balances, Cubis® II MCA
Toimittaja: Sartorius Balances
MCA32202P2S000 MCA70201S2S000 MCA50201S2S000 MCA36201S2S000 MCA36201P2S000 MCA20201S2S000 MCA70200S2S000 MCA36200S2S000 MCA32202P2099 MCA70201S2099 MCA50201S2099 MCA36201S2099 MCA36201P2099 MCA20201S2099 MCA70200S2099 MCA36200S2099
SARBMCA32202P2S000EA 16710 EUR
SARBMCA32202P2S000 SARBMCA70201S2S000 SARBMCA50201S2S000 SARBMCA36201S2S000 SARBMCA36201P2S000 SARBMCA20201S2S000 SARBMCA70200S2S000 SARBMCA36200S2S000 SARBMCA32202P2099 SARBMCA70201S2099 SARBMCA50201S2099 SARBMCA36201S2099 SARBMCA36201P2099 SARBMCA20201S2099 SARBMCA70200S2099 SARBMCA36200S2099
High-capacity toploading laboratory balances, Cubis® II MCA
Vaa'at
Cubis® II MCA high-capacity toploading balances with advanced user interface offer a maximum load between 11,2 and 70,2 kg and readability between 0,1 and 1 g provide the ideal model for every application.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringentMore Product Information