Analytical laboratory balances, Cubis® II MCA
Toimittaja: Sartorius Balances
MCA524S2S00U99 MCA524P2S00U99 MCA324S2S00U99 MCA324P2S00U99 MCA224S2S00U99 MCA124S2S00U99 MCA524S2S00A99 MCA524P2S00A99 MCA324S2S00A99 MCA324P2S00A99 MCA224S2S00A99 MCA124S2S00A99 MCA524S2S00I99 MCA524P2S00I99 MCA324S2S00I99 MCA324P2S00I99 MCA224S2S00I99 MCA124S2S00I99 MCA324P2S00-I MCA524S2S00-U MCA524P2S00-U MCA324S2S00-U MCA324P2S00-U MCA224S2S00-U MCA124S2S00-U MCA524S2S00-A MCA524P2S00-A MCA324S2S00-A MCA324P2S00-A MCA124S2S00-A MCA524S2S00-I MCA524P2S00-I MCA324S2S00-I MCA224S2S00-I MCA124S2S00-I MCA224S-2S00-A
SARBMCA524S2S00U99EA 17050 EUR
SARBMCA524S2S00U99 SARBMCA524P2S00U99 SARBMCA324S2S00U99 SARBMCA324P2S00U99 SARBMCA224S2S00U99 SARBMCA124S2S00U99 SARBMCA524S2S00A99 SARBMCA524P2S00A99 SARBMCA324S2S00A99 SARBMCA324P2S00A99 SARBMCA224S2S00A99 SARBMCA124S2S00A99 SARBMCA524S2S00I99 SARBMCA524P2S00I99 SARBMCA324S2S00I99 SARBMCA324P2S00I99 SARBMCA224S2S00I99 SARBMCA124S2S00I99 SARBMCA324P2S00-I SARBMCA524S2S00-U SARBMCA524P2S00-U SARBMCA324S2S00-U SARBMCA324P2S00-U SARBMCA224S2S00-U SARBMCA124S2S00-U SARBMCA524S2S00-A SARBMCA524P2S00-A SARBMCA324S2S00-A SARBMCA324P2S00-A SARBMCA124S2S00-A SARBMCA524S2S00-I SARBMCA524P2S00-I SARBMCA324S2S00-I SARBMCA224S2S00-I SARBMCA124S2S00-I 611-5264
Analytical laboratory balances, Cubis® II MCA
Vaa'at
Cubis® II MCA analytical balances with advanced user interface offer maximum weighing capacity between 120 and 520 g in combination with readability of 0.1 mg.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliverMore Product Information