Cubis® II MCA 125S Ultra-High Resolution Semi-Micro Balances
Toimittaja: Sartorius Balances
MCA125S3S0M MCA125S3CE3H MCA125S3S013H MCA125S3CE99I MCA125S3S013I MCA125S3CE23 MCA125S3S034H MCA125S3S034M MCA125S3CE123M MCA125S3S024 MCA125S3CE24M MCA125S3CE123 MCA125S3CE13H MCA125S3S024M MCA125S3CE1I MCA125S3S013M MCA125S3CE34H MCA125S3CE234M MCA125S3S012 MCA125S3S03 MCA125S3CE24I MCA125S3CE4M MCA125S3CE12 MCA125S3CE12I MCA125S3S023H MCA125S3CE1M MCA125S3CE3I MCA125S3S0234 MCA125S3CE2H MCA125S3S0234M MCA125S3S02I MCA125S3S014M MCA125S3CE123I MCA125S3CE3M MCA125S3S034 MCA125S3S0123I MCA125S3CE12M MCA125S3S04H MCA125S3CE14M MCA125S3S034I MCA125S3S099I MCA125S3S02H MCA125S3CE14I MCA125S3S013 MCA125S3CE34I MCA125S3S014 MCA125S3S0234H MCA125S3S023I MCA125S3S012I MCA125S3S014H MCA125S3CE2M MCA125S3CE23I MCA125S3CEM MCA125S3CE124 MCA125S3CE34 MCA125S3S01I MCA125S3S02 MCA125S3CE99M MCA125S3S02M MCA125S3CE24 MCA125S3CE23M MCA125S3S03I MCA125S3CE2I MCA125S3CE234I MCA125S3CEI MCA125S3CE4I MCA125S3CE24H MCA125S3S03H MCA125S3CE123H MCA125S3CE14H MCA125S3S023M MCA125S3S0124M MCA125S3S0124 MCA125S3CE2 MCA125S3CE234H MCA125S3S012M MCA125S3CE4 MCA125S3S03M MCA125S3CE124I MCA125S3S024H MCA125S3S0124I MCA125S3S0234I MCA125S3S0124H MCA125S3CE14 MCA125S3CE12H MCA125S3S04M MCA125S3CE13M MCA125S3S012H MCA125S3S024I MCA125S3CE23H MCA125S3CE234 MCA125S3CE13I MCA125S3S0123 MCA125S3S014I MCA125S3S0123M MCA125S3CE4H MCA125S3CE34M MCA125S3S023 MCA125S3S01M MCA125S3S099M MCA125S3CE124M MCA125S3CE124H MCA125S3CE13 MCA125S3S04 MCA125S3S0123H MCA125S3S04I MCA125S3S0I MCA125S3CE3
SARBMCA125S3S0MEA 18010 EUR
SARBMCA125S3S0M SARBMCA125S3CE3H SARBMCA125S3S013H SARBMCA125S3CE99I SARBMCA125S3S013I SARBMCA125S3CE23 SARBMCA125S3S034H SARBMCA125S3S034M SARBMCA125S3CE123M SARBMCA125S3S024 SARBMCA125S3CE24M SARBMCA125S3CE123 SARBMCA125S3CE13H SARBMCA125S3S024M SARBMCA125S3CE1I SARBMCA125S3S013M SARBMCA125S3CE34H SARBMCA125S3CE234M SARBMCA125S3S012 SARBMCA125S3S03 SARBMCA125S3CE24I SARBMCA125S3CE4M SARBMCA125S3CE12 SARBMCA125S3CE12I SARBMCA125S3S023H SARBMCA125S3CE1M SARBMCA125S3CE3I SARBMCA125S3S0234 SARBMCA125S3CE2H SARBMCA125S3S0234M SARBMCA125S3S02I SARBMCA125S3S014M SARBMCA125S3CE123I SARBMCA125S3CE3M SARBMCA125S3S034 SARBMCA125S3S0123I SARBMCA125S3CE12M SARBMCA125S3S04H SARBMCA125S3CE14M SARBMCA125S3S034I SARBMCA125S3S099I SARBMCA125S3S02H SARBMCA125S3CE14I SARBMCA125S3S013 SARBMCA125S3CE34I SARBMCA125S3S014 SARBMCA125S3S0234H SARBMCA125S3S023I SARBMCA125S3S012I SARBMCA125S3S014H SARBMCA125S3CE2M SARBMCA125S3CE23I SARBMCA125S3CEM SARBMCA125S3CE124 SARBMCA125S3CE34 SARBMCA125S3S01I SARBMCA125S3S02 SARBMCA125S3CE99M SARBMCA125S3S02M SARBMCA125S3CE24 SARBMCA125S3CE23M SARBMCA125S3S03I SARBMCA125S3CE2I SARBMCA125S3CE234I SARBMCA125S3CEI SARBMCA125S3CE4I SARBMCA125S3CE24H SARBMCA125S3S03H SARBMCA125S3CE123H SARBMCA125S3CE14H SARBMCA125S3S023M SARBMCA125S3S0124M SARBMCA125S3S0124 SARBMCA125S3CE2 SARBMCA125S3CE234H SARBMCA125S3S012M SARBMCA125S3CE4 SARBMCA125S3S03M SARBMCA125S3CE124I SARBMCA125S3S024H SARBMCA125S3S0124I SARBMCA125S3S0234I SARBMCA125S3S0124H SARBMCA125S3CE14 SARBMCA125S3CE12H SARBMCA125S3S04M SARBMCA125S3CE13M SARBMCA125S3S012H SARBMCA125S3S024I SARBMCA125S3CE23H SARBMCA125S3CE234 SARBMCA125S3CE13I SARBMCA125S3S0123 SARBMCA125S3S014I SARBMCA125S3S0123M SARBMCA125S3CE4H SARBMCA125S3CE34M SARBMCA125S3S023 SARBMCA125S3S01M SARBMCA125S3S099M SARBMCA125S3CE124M SARBMCA125S3CE124H SARBMCA125S3CE13 SARBMCA125S3S04 SARBMCA125S3S0123H SARBMCA125S3S04I SARBMCA125S3S0I SARBMCA125S3CE3
Cubis® II MCA 125S Ultra-High Resolution Semi-Micro Balances
Vaa'at
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCA display - 7"More Product Information