X-ray and metal detectable powder funnels
Toimittaja: Sampling Systems
M1051B-200
SAMPM1051B-200EA 59.6 EUR
SAMPM1051B-200
X-ray and metal detectable powder funnels
Suppilot Jauhesuppilot
X-ray and metal detectable powder funnels are ideal for both laboratory and production work. The bright blue colour makes the bottles easy to spot if they are left in the product.
  • Robust PP construction
  • Non sterile
  • PP gives excellent chemical resistance to many products
  • Full batch traceability
To ensure the cleanliness of the funnels, they are manufactured and packed in a cleanroom. The robust nature of these funnels means that they can give many years of service or for more demanding applications. They can be used as a single use disposable productMore Product Information