Mini-Drums and Light Duty Drums (No Toogles)
Toimittaja: Sampling Systems
A446-15 A446-1 A446-10 A446-5 A466-20 A466-25 A466-30 A466-50 A446-2 A446-6 A454-25 A454-20 A454-30 A454-50 A446-8 A446-12 A466-100 A466-75
SAMPA446-15EA 1670 EUR
SAMPA446-15 SAMPA446-1 SAMPA446-10 SAMPA446-5 SAMPA466-20 SAMPA466-25 SAMPA466-30 SAMPA466-50 SAMPA446-2 SAMPA446-6 SAMPA454-25 SAMPA454-20 SAMPA454-30 SAMPA454-50 SAMPA446-8 SAMPA446-12 SAMPA466-100 SAMPA466-75
Mini-Drums and Light Duty Drums (No Toogles)
Tynnyrit, tynnyrit, rummut
Made of stainless steel.
  • Light duty construction
  • With crevice free interior
  • Flat bottom with chime ring to give added protection
  • With lid
Multi purpose containers.
Varoitus: If you need a watertight container please ask for drum with toggles.