Bottles, Aluminium, with tamper evident lid
Toimittaja: Sampling Systems
A179-50 A179-100 A179-250 A179-500 A179-1000
SAMPA179-50EA 4.3 EUR
SAMPA179-50 SAMPA179-100 SAMPA179-250 SAMPA179-500 SAMPA179-1000
Bottles, Aluminium, with tamper evident lid
Pullot Metallipullot
Aluminium bottles are very popular in general laboratory and production work.
  • Great for storing samples
  • Available in a wide range of sizes
  • Two part sealing system to ensure water tightness
  • Fast delivery
These aluminium bottles have an intuitive sealing system which makes them water tight and excellent for storing samples.