Volumetric scoops, USP Class VI, double-bagged, ULTRA
Toimittaja: Sampling Systems
9286H-1000S 9286H-100S 9286H-150S 9286H-250S 9286H-25S 9286H-500S 9286H-50S
SAMP9286H-1000SEA 338 EUR
SAMP9286H-1000S SAMP9286H-100S SAMP9286H-150S SAMP9286H-250S SAMP9286H-25S SAMP9286H-500S SAMP9286H-50S
Volumetric scoops, USP Class VI, double-bagged, ULTRA
Kauhat
The SteriWare Ultra range is the premium range from Sampling Systems. These premium sterile scoops are double bagged and are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
  • Sterilised by gamma irradiation
  • Available in a range of sizes from 25 to 1000 ml
The scoops all have an ergonomic handle and are easy to use. The volumetric shape means specific volumes can be quickly measured. The flat base means they also sitMore Product Information