Sample Spoons, Disposable, SteriWare®
Toimittaja: Sampling Systems
8088A-10S 8088A-03S 8088A-03 8088A-10 8088R-10S 8088B-10S 8088B-10
SAMP8088A-10SEA 154 EUR
SAMP8088A-10S SAMP8088A-03S SAMP8088A-03 SAMP8088A-10 SAMP8088R-10S SAMP8088B-10S SAMP8088B-10
Sample Spoons, Disposable, SteriWare®
Lusikat
These disposable, sterile sample spoons are ideal for dispensing and sampling small quantities. Manufactured from food grade polystyrene (PS) or polyproylene (PP).
  • Moulded and packed in a cleanroom
  • Available in white, red and blue
  • Made from FDA and EU conforming polymers
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
Full batch traceability.
Pakkaus: Individually bagged.