Näytteenottimet, Slot samplers, kertakäyttöiset, SteriWare®
Toimittaja: Sampling Systems
8073H-1000 8073H-500 8073H-1000S 8073H-500S 8073J-1000S 8073J-1000 8073H-300
300-0041EA 477 EUR
300-0041 300-0043 300-0042 300-0044 300-0217 SAMP8073J-1000 SAMP8073H-300
Näytteenottimet, Slot samplers, kertakäyttöiset, SteriWare®
Näytteenottimet Näytteenottimet, kertakäyttöiset
These cross sectional multilevel disposable powder samplers are designed for powders. The robust HDPE construction make this to be used in huge number of testing applications.
  • BSE/TSE-free
  • Full batch traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Available as gamma irradiated sterile (SAL 10-6)
Can take a composite from the top, middle and bottom of the powder bed – Moulded and packed in a cleanroom to ensure cleanliness.
Ideal for taking a multilevel, cross sectional sample from powders and granules. Insert the sampler and rotate theMore Product Information