Tablet counting triangles, SteriWare®
Toimittaja: Sampling Systems
8066A-1 8066A-1S
SAMP8066A-1EA 222 EUR
SAMP8066A-1 SAMP8066A-1S
Tablet counting triangles, SteriWare®
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation
Sertifikaatit: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Pakkaus: Individually bagged in a heat-sealed laminate bag.