Kauhat, kertakäyttöiset, PharmaScoop®, SteriWare®
Toimittaja: Sampling Systems
8045A-60 8045A-60S 8045A-125 8045A-125S 8045A-250 8045A-2500 8045A-30 8045A-30S 8045B-2500S 8045B-60S 8045H-1000 8045H-125 8045H-500 8045A-2500S 8045A-1000 8045R-60S 8045B-60 8045H-125S 8045H-60S 8045H-250 8045H-60 8045R-125S 8045R-30S 8045R-500_U 8045R-60 8045B-125 8045B-125S 8045B-250 8045B-250S 8045H-30 8045P-2500 8045P-60 8045P-60S 8045R-1000 8045B-500S 8045P-250 8045P-250S 8045A-250S 8045H-250S 8045P-125 8045P-125S 8045P-30 8045P-30S 8045R-125 8045R-250 8045R-30 8045R-250S 8045A-1000S 8045A-500 8045A-500S 8045H-1000S 8045H-500S 8045P-1000 8045P-1000S 8045P-2500S 8045P-500 8045P-500S 8045R-1000S 8045R-2500 8045R-2500S 8045R-500 8045R-500S
231-0420EA 211 EUR
231-0420 231-0421 231-0375 231-0376 231-0377 231-0378 231-0381 231-0382 231-0423 231-0424 SAMP8045H-1000 SAMP8045H-125 SAMP8045H-500 231-0379 231-0449 231-0454 231-0595 SAMP8045H-125S SAMP8045H-60S SAMP8045H-250 SAMP8045H-60 SAMP8045R-125S SAMP8045R-30S 231-0596 231-0594 SAMP8045B-125 SAMP8045B-125S SAMP8045B-250 SAMP8045B-250S SAMP8045H-30 SAMP8045P-2500 SAMP8045P-60 SAMP8045P-60S SAMP8045R-1000 SAMP8045B-500S SAMP8045P-250 SAMP8045P-250S 231-0380 SAMP8045H-250S 300-0522 300-0523 300-0524 300-0525 300-0526 300-0528 300-0529 SAMP8045R-250S SAMP8045A-1000S SAMP8045A-500 SAMP8045A-500S SAMP8045H-1000S SAMP8045H-500S SAMP8045P-1000 SAMP8045P-1000S SAMP8045P-2500S SAMP8045P-500 SAMP8045P-500S SAMP8045R-1000S SAMP8045R-2500 SAMP8045R-2500S SAMP8045R-500 SAMP8045R-500S
Kauhat, kertakäyttöiset, PharmaScoop®, SteriWare®
Kauhat
Single-use plastic scoops for hygienic applications. These PharmaScoops are ideal for both sampling and dispensing powders and granules.
  • FDA conforming materials
  • Free from BSE/TSE and crevices
  • Full batch traceability
  • Sterile scoops are gamma irradiated (SAL 10⁻⁶)
  • Available in red, white and blue
  • Coloured versions are easy to spot if they are left in the powder by mistake
Useful alternative to traditional stainless steel scoops. They are moulded and packed in a Class 10 000 cleanroom environment. Each scoop is individually wrapped so can be usedMore Product Information