Stainless Chain
Toimittaja: Sampling Systems
5070S-50
SAMP5070S-50EA 978 EUR
SAMP5070S-50
Stainless Chain
Kourat Yleiskäyttöiset puristimet
Our popular range of accessories are essential when sampling with Sub Samplers (Bomb Sampler) and bailers. Stainless chain provides great chemical resistance.