Cohesive Slot Samplers
Toimittaja: Sampling Systems
2210A-600 2210A-1000 2210A-1500
SAMP2210A-600EA 1840 EUR
SAMP2210A-600 SAMP2210A-1000 SAMP2210A-1500
Cohesive Slot Samplers
Näytteenottimet Kiinteiden Aineiden Näytteenottimet
The cohesive slot sampler is designed for taking cross sectional samples of cohesive powders.
  • Made from 316L stainless steel
  • Can be completely dismantled for quick and thorough cleaning
  • Available from stock
  • Fast delivery
The cohesive slot sampler is excellent for taking cross sectional samples from powders with poor flow characteristics. Upon rotating the device, the scraper pushes powder into the sampler. When finished, the sampler can be compeltely dismantled for quick and easy cleaning.