Slot sampler with bottle adapter
Toimittaja: Sampling Systems
1210B-600
SAMP1210B-600EA 1030 EUR
SAMP1210B-600
Slot sampler with bottle adapter
Näytteenottimet Kiinteiden Aineiden Näytteenottimet
The slot sampler is perfect for taking large volume, cross sectional samples from containers, big bags, bins and so on.
  • Great for large volume sampling
  • Robust stainless steel construction
  • Ideal for quality control in the food, chemical, and pharmaceutical industries
  • Double wall design means powder cannot enter the sampler until open
Made from 316L stainless steel, the slot sampler is available in a range of sizes.