Näytteenottimet, PowderThief
Toimittaja: Sampling Systems
1030C-600 1030CT-10 1030CT-25 1030D-1000 1030D-1500 1030AT-050 1030A-600 1030AT-075 1030AT-100 1030A-1500 1030A-1000 1030A-300 1030AT-025 1030DT-50 1030DT-100 1030DT-20 1030DT-25 1030B-600 1030BT-2 1030BT-3 1030B-1000 1030BT-5 1030B-1500 1030D-600 1030C-1000 1030C-1500 1030D-2000 1030AT-150 1030BT-10 1030BT-8
SAMP1030C-600EA 875 EUR
SAMP1030C-600 SAMP1030CT-10 SAMP1030CT-25 SAMP1030D-1000 SAMP1030D-1500 SAMP1030AT-050 SAMP1030A-600 SAMP1030AT-075 SAMP1030AT-100 SAMP1030A-1500 SAMP1030A-1000 SAMP1030A-300 SAMP1030AT-025 SAMP1030DT-50 SAMP1030DT-100 SAMP1030DT-20 SAMP1030DT-25 SAMP1030B-600 SAMP1030BT-2 SAMP1030BT-3 SAMP1030B-1000 SAMP1030BT-5 SAMP1030B-1500 SAMP1030D-600 SAMP1030C-1000 SAMP1030C-1500 SAMP1030D-2000 SAMP1030AT-150 SAMP1030BT-10 SAMP1030BT-8
Näytteenottimet, PowderThief
Näytteenottimet Kiinteiden Aineiden Näytteenottimet
These PowderThief samplers are suitable for sampling free flowing powders and granules.
  • Made of high quaility 316 stainless steel
  • PTFE used for clutch mechanism
  • Single point sampler
A range of tips are available so that a single sampler can be used to accurately sample a range of different volumes.