Exact volume transfer pipettes, Samco™, Thermo Scientific
Toimittaja: SAMCO
787NL 966NL 191NL 783NL 784NL 785NL 786NL 788NL 930NL 941NL 967NL
SAMC787NLEA 0 EUR
SAMC787NL SAMC966NL SAMC191NL SAMC783NL SAMC784NL SAMC785NL SAMC786NL SAMC788NL SAMC930NL SAMC941NL SAMC967NL
Exact volume transfer pipettes, Samco™, Thermo Scientific
Pipetit ja pipetintäyttäjät Pasteur-/siirtopipetit
The exact volume transfer pipettes are used for dispensing reagents of small samples.
  • Polyethylene
  • Non sterile
  • Shatterproof
  • Ideal for home and POC (Point of Care) testing
  • OEM applications
Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage. Useful in clinical laboratories. Custom design options available to meet your specifications.