Transfer pipettes, Samco™ Extra Long
Toimittaja: SAMCO
262-1S 262-20S 262-10S 263 263-1S
SAMC262-1SEA 0 EUR
SAMC262-1S SAMC262-20S SAMC262-10S SAMC263 SAMC263-1S
Transfer pipettes, Samco™ Extra Long
Pipetit ja pipetintäyttäjät Pasteur-/siirtopipetit
These extra long PE pipettes are ideal for safe and easy sampling from large containers.
  • Excellent for tissue culture applications
  • General purpose pipettes are ideal for routine sampling of serum and plasma
  • Well suited for blood culture and urine analysis
Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.