Foetal bovine serum (FBS), US origin
Toimittaja: BIOWEST
S1520-100 S1520-500 S152H-100 S152H-500
S1520-100EA 0 EUR
S1520-100 S1520-500 S152H-100 S152H-500
Foetal bovine serum (FBS), US origin
Seerumit
Sterile filtered foetal bovine serum (FBS) derived from clotted whole blood aseptically collected from foetus via cardiac puncture. Each manufactured batch is rigorously controlled, from the collection of serum through to final packaging.
  • USA origin
  • High quality serum, rigorously tested
  • Full traceability and security
Sertifikaatit: Biowest is a certified ISO 9001:2008 company. Registered by the French Ministry of Agriculture (EC regulation No. 1069/2009) under agreement no. FR 49.231.001 for the production of animal sera.