HYGEN™ Microfibre Cloths
Toimittaja: Rubbermaid Commercial Products
FGQ61000YL00 FGQ62000BL00 FGQ62000GR00 FGQ62000RD00
RUBBFGQ61000YL00EA 6.4 EUR
RUBBFGQ61000YL00 RUBBFGQ62000BL00 RUBBFGQ62000GR00 RUBBFGQ62000RD00
HYGEN™ Microfibre Cloths
Pyyhkeet, liinat ja sienet Uudelleenkäytettävät puhdistusliinat Liinat yleiskäyttöön
Reduce the spread of germs with quality microfibre cloths that provide excellent cleaning performance.
  • Durable for up to 500 launderings (up to 200 with bleach)
  • Patented zig-zag scrubbers make cleanup quick and easy
  • Removes 99,7% or more of tested virus and bacteria
  • Easy to clean; bleach safe in hot and cold water
  • High-quality, 16-split microfibre penetrates deeper for a superior clean
  • Facilitate colour-coding implementation by assigning a coloured pad to a specific area
  • Ideal for dusting and wet cleaning
  • Compatible with common disinfectants: Quad,