Flex™ Dispensers
Toimittaja: Rubbermaid Commercial Products
3486589 3486590 3486591 3486592
RUBB3486589EA 22.6 EUR
RUBB3486589 RUBB3486590 RUBB3486591 RUBB3486592
Flex™ Dispensers
Saippua- ja voide- ja saippua-annostelijat
Tailored hygiene for complete confidence. Flexible manual dispenser with the widest range of foam and lotion refills, and adjustable dose sizes.
  • Adjustable dose size for effective cost control (0,4, 0,65 and 1 ml)
  • Unique dispenser design allows foam and lotion refills in the same dispenser
  • Broadest formulation range, including hand washes, body washes, and hand sanitisers
  • Antimicrobial pushbar is protected with BioMaster technology
  • Highest hand wash yield; up to 3250 doses per refill
  • Hygienically sealed refills keep soap and hand sanitisers fresh and avoid