Instrument trays, rectangular
Toimittaja: RSG
701.252 701.220 701.242 701.235 701.230 701.228 701.226 701.222 701.217 701.224
RSGA701.252EA 280 EUR
RSGA701.252 RSGA701.220 RSGA701.242 RSGA701.235 RSGA701.230 RSGA701.228 RSGA701.226 RSGA701.222 RSGA701.217 RSGA701.224
Instrument trays, rectangular
Haihdutusmaljat/Tarjottimet/Altaat Instrumenttialustat/-astiat
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance