Coated tweezers
Toimittaja: RSG
411.130 411.105 413.200 414.200 414.160 414.145 414.130 414.105 413.160 413.145 413.130 413.105 411.200 411.160 411.145 414.115
RSGA411.130EA 98.8 EUR
RSGA411.130 RSGA411.105 RSGA413.200 RSGA414.200 RSGA414.160 RSGA414.145 RSGA414.130 RSGA414.105 RSGA413.160 RSGA413.145 RSGA413.130 RSGA413.105 RSGA411.200 RSGA411.160 RSGA411.145 RSGA414.115
Coated tweezers
Pinsetit
These standard tweezers are made of stainless steel (Remanit® 4301) and feature a PTFE coating.