Support rings
Toimittaja: RSG
280.050 280.070 280.100 280.130 280.160 281.050 281.070 281.100 281.130 281.160 283.050 283.070 283.100 283.130 283.160 284.050 284.070 284.100 284.130 284.160
241-0317EA 21.7 EUR
241-0317 241-0318 RSGA280.100 241-0319 RSGA280.160 241-0320 241-0321 241-0322 241-0323 RSGA281.160 241-0324 241-0325 RSGA283.100 241-0326 RSGA283.160 RSGA284.050 241-0327 RSGA284.100 RSGA284.130 RSGA284.160
Support rings
Statiivirenkaat
These heavy-duty support rings are made from stainless steel.
  • 8 mm rod
  • Available with rubber covered rings