Microarray hybridisation solution v2.0
Toimittaja: Cytiva
RPK0325
RPK0325EA 198 EUR
RPK0325
Microarray hybridisation solution v2.0
Microarray -reagenssit
Microarray hybridisation solution version 2.0, for highly reproducible microarray results.
  • Includes a blend of proprietary components to provide highly reproducible, high-quality automated and manual DNA microarray hybridisations
  • Hybridisation rate enhancers improve the detection of rare targets
  • Proprietary blocking agents minimize non-specific background
DNA microarrays can be used to measure the expression levels of large numbers of genes simultaneously (gene expression profiling), or to genotype multiple areas of a genome (SNP arrays), with variable coverage or resolution depending how many DNA 'spots' cover those areas.