Tetrabutylammonium hydroxide 40% vesiliuos

Toimittaja: Roth Carl
Vaara

5026.1
ROTH5026.1EA 64 EUR
ROTH5026.1
Tetrabutylammonium hydroxide 40% vesiliuos
Tetrabutylammonium hydroxide
Kaava: C₁₆H₃₇NO
Storage Temperature: Jääkaappi
MDL numero: MFCD00009425
CAS-numero: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR