Acacetin

Toimittaja: Roth Carl

5010.1
ROTH5010.1EA 0 EUR
ROTH5010.1
Acacetin
Acacetin
Kaava: C₁₆H₁₂O₅
MDL numero: MFCD00016936
CAS-numero: 480-44-4
EINECS: 207-552-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR