Hoffmann pinch cocks, Rotilabo®
Toimittaja: Roth Carl
1585.1 1586.1 1587.1 1588.1 1589.1
ROTH1585.1EA 0 EUR
ROTH1585.1 ROTH1586.1 ROTH1587.1 ROTH1588.1 ROTH1589.1
Hoffmann pinch cocks, Rotilabo®
Kourat Letkupuristimet
Rotilabo®-tube clamping screws made of chrome-plated metal for fine regulation.
  • Tube clamping screws according to Hoffmann
  • Hinged bottom bar can be swing open