5-bromi-4-kloori-3-indolyyli-beta-D-galaktopyranosidi, X-Gal

Toimittaja: Rockland Immunochemicals

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

XGAL-0100
ROCKXGAL-0100EA 227 EUR
ROCKXGAL-0100
5-bromi-4-kloori-3-indolyyli-beta-D-galaktopyranosidi, X-Gal
5-bromi-4-kloori-3-indolyyli-beta-D-galaktopyranosidi, X-Gal
Kaava: C₁₄H₁₅BrClNO₆
Sulamispiste: 237…239 °C
Storage Temperature: Pakastin
MDL numero: MFCD00005666
CAS-numero: 7240-90-6
EINECS: 230-640-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR