Anti-IgA/IgG/IgM Goat Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
710-101-130
ROCK710-101-130EA 325 EUR
ROCK710-101-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Vasta-aineet
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000;

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR