Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Rockland Immunochemicals
704-403-002
ROCK704-403-002EA 396 EUR
ROCK704-403-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Recommended Dilutions: ELISA: 1:85000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR