Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 680)

Toimittaja: Rockland Immunochemicals
612-144-002-05
ROCK612-144-002-05EA 690 EUR
ROCK612-144-002-05
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 680)
Vasta-aineet
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR