Anti-IgG (H+L) Chicken Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
609-9102
ROCK609-9102EA 290 EUR
ROCK609-9102
Anti-IgG (H+L) Chicken Polyclonal Antibody
Vasta-aineet
Secondary Chicken Anti-IgG (H&L) Reacts with Human

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR