Anti-IgG F(c) Goat Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
609-1103
ROCK609-1103EA 421 EUR
ROCK609-1103
Anti-IgG F(c) Goat Polyclonal Antibody
Vasta-aineet
Human IgG F(c) Antibody is suitable for immunoassays where specificity to the immunoglobulin Fc region is desired. Antibody has been tested by ELISA, western blot immunoblot, and immunohistochemisty.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:10000 - 1:50000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR