Anti-IgG F(c) Rabbit Polyclonal Antibody (Texas Red®)

Toimittaja: Rockland Immunochemicals
608-4903
ROCK608-4903EA 384 EUR
ROCK608-4903
Anti-IgG F(c) Rabbit Polyclonal Antibody (Texas Red®)
Vasta-aineet
Secondary Rabbit Anti-IgG F(c) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR