Anti-HIST4H4 Rabbit Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
600-401-J02
ROCK600-401-J02EA 856 EUR
ROCK600-401-J02
Anti-HIST4H4 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:50; Western Blot: 1:2000; Immunochemistry: 1:50; Other Dilution: Dot Blot 1:1000

Type: Primary
Antigen: HIST4H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR