Anti-HIST2H3C Rabbit Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
600-401-I89
ROCK600-401-I89EA 856 EUR
ROCK600-401-I89
Anti-HIST2H3C Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Other Dilution: Dot Blot 1:1000

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR