Anti-COL1A1 Rabbit Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
600-401-D19
ROCK600-401-D19EA 980 EUR
ROCK600-401-D19
Anti-COL1A1 Rabbit Polyclonal Antibody
Vasta-aineet
Anti-Collagen I, alpha 1 propeptide Antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to collagen I alpha I.

Recommended Dilutions: ELISA: 1:5000-1:50000; Western Blot: 1:1000; Immunochemistry: 1:100; Immunoprecipitation: 1:100

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR