Anti-AKT1 Rabbit Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
600-401-268
ROCK600-401-268EA 781 EUR
ROCK600-401-268
Anti-AKT1 Rabbit Polyclonal Antibody
Vasta-aineet
This antibody is phospho specific for pS473 and is suitable for western blotting, immunochemistry (formalin-fixed paraffin-embedded sections), immunofluorescence and ELISA. By immunoblot a single band of the expected apparent molecular weight is obse

Recommended Dilutions: ELISA: 1:15000 - 1:60,000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:100 - 1:500;

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR