Anti-RBC Rabbit Polyclonal Antibody
Toimittaja: Rockland Immunochemicals
213-4139
ROCK213-4139EA 709 EUR
ROCK213-4139
Anti-RBC Rabbit Polyclonal Antibody
Vasta-aineet
Anti-Sheep Red Blood Cell Antibody may be used in hemagglutination assays. Anti-SHEEP Red Blood Cell Antibody is used to sensitize erythrocytes and quantitate agglutination. Haemagglutination assay or HA is a method of quantification for viruses or bacteria by hemagglutination. Some viral families and many bacteria have envelope or surface proteins which are able to agglutinate (stick to) human or animal red blood cells (RBC) and bind to N-acetylneuraminic acid. As each of the agglutinating molecule attaches to multiple RBCs, a lattice-structure will form. Normally, a virus dilution (e.g. 2-More Product Information