Anti-ODF2 Rabbit Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
200-401-C43S
ROCK200-401-C43SEA 226 EUR
ROCK200-401-C43S
Anti-ODF2 Rabbit Polyclonal Antibody
Vasta-aineet
This Protein-A purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 93 kDa in size corresponding to Cenexin-1 by western blotting.

Recommended Dilutions: ELISA: 1:500-1:10000; Western Blot: 1:500-1:2000;

Type: Primary
Antigen: ODF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR