Anti-GFAP Rabbit Polyclonal Antibody
Toimittaja: Rockland Immunochemicals
100-401-D60
ROCK100-401-D60EA 861 EUR
ROCK100-401-D60
Anti-GFAP Rabbit Polyclonal Antibody
Vasta-aineet
Glial Fibrillary Acidic Protein (GFAP) was discovered by Amico Bignami and co-workers as a major fibrous protein of multiple sclerosis plaques. It was subsequently found to be a member of the 10nm or intermediate filament (IF) family, specifically the IF family Class III, which also includes peripherin, desmin and vimentin. GFAP is strongly and specifically expressed in astrocytes and certain other astroglia in the CNS, in satellite cells, peripheral ganglia, and in non-myelinating Schwann cells in peripheral nerves. In many damage and disease states GFAP expression is heavily upregulated inMore Product Information