Anti-CYTIP Rabbit Polyclonal Antibody

Toimittaja: Rockland Immunochemicals
100-401-A23
ROCK100-401-A23EA 821 EUR
ROCK100-401-A23
Anti-CYTIP Rabbit Polyclonal Antibody
Vasta-aineet
This antibody has been tested for use in ELISA and immunoprecipitation.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 40 kDa in size corresponding to Cybr by western blotting after IP.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000 - 1:10000; Immunoprecipitation: 1:100
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: CYTIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR