VitraPOR® Glass Filter Discs, Biplane
Toimittaja: ROBU GLASFILTER
16104 16100 16101 16301 16303 16302 16304 16309 16400 16401 16402 16403 16404 16409 16500 16501 16502 16503 16600 16504 16802 16601 16602 16603 16604 16609 16700 16701 16702 16703 16704 16709 16801 16803 16804 16809 16900 16901 16902 16903 16904 16909 16509 16800 16200 16109 16110 16111 16112 16113 16114 16119 16120 16121 16122 16123 16103 16129 16102 16201 16202 16203 16204 16209 16250 16251 16252 16253 16254 16259 16300 16124 61803 62004 62003 62002 62001 62000 61904 61903 61902 61901 61900 61301 62102 61303 62303 61300 61302 62304 62601 62802 62801 62800 62704 62703 62702 62701 62700 62604 62804 62602 62900 62600 62504 62503 62502 62501 62500 62404 62403 62402 62401 62400 62603 63502 63804 63803 63802 63801 63800 62803 63503 63501 63500 63004 63003 63002 63001 63000 62904 62903 62902 62901 63504 61504 61801 61800 61704 61703 61702 61701 61700 61604 61603 61602 61600 61804 61503 61502 61501 61500 61404 61403 61402 61401 61400 61304 61601 62100 62302 62301 62300 62204 62203 62202 62201 62200 62104 62103 61802 62101 16105 16115 16125 16205 16255 16305 16405 16505 16605 16705 16805 16905 16999 6130F 6140F 6150F 6160F 6170F 6180F 6190F 6200F 1610F 1611F 1612F 1620F 1625F 1630F 1640F 1650F 1660F 1670F 1680F 1690F
ROBU16104EA 88.4 EUR
ROBU16104 ROBU16100 ROBU16101 ROBU16301 ROBU16303 ROBU16302 ROBU16304 ROBU16309 ROBU16400 ROBU16401 ROBU16402 ROBU16403 ROBU16404 ROBU16409 ROBU16500 ROBU16501 ROBU16502 ROBU16503 ROBU16600 ROBU16504 ROBU16802 ROBU16601 ROBU16602 ROBU16603 ROBU16604 ROBU16609 ROBU16700 ROBU16701 ROBU16702 ROBU16703 ROBU16704 ROBU16709 ROBU16801 ROBU16803 ROBU16804 ROBU16809 ROBU16900 ROBU16901 ROBU16902 ROBU16903 ROBU16904 ROBU16909 ROBU16509 ROBU16800 ROBU16200 ROBU16109 ROBU16110 ROBU16111 ROBU16112 ROBU16113 ROBU16114 ROBU16119 ROBU16120 ROBU16121 ROBU16122 ROBU16123 ROBU16103 ROBU16129 ROBU16102 ROBU16201 ROBU16202 ROBU16203 ROBU16204 ROBU16209 ROBU16250 ROBU16251 ROBU16252 ROBU16253 ROBU16254 ROBU16259 ROBU16300 ROBU16124 ROBU61803 ROBU62004 ROBU62003 ROBU62002 ROBU62001 ROBU62000 ROBU61904 ROBU61903 ROBU61902 ROBU61901 ROBU61900 ROBU61301 ROBU62102 ROBU61303 ROBU62303 ROBU61300 ROBU61302 ROBU62304 ROBU62601 ROBU62802 ROBU62801 ROBU62800 ROBU62704 ROBU62703 ROBU62702 ROBU62701 ROBU62700 ROBU62604 ROBU62804 ROBU62602 ROBU62900 ROBU62600 ROBU62504 ROBU62503 ROBU62502 ROBU62501 ROBU62500 ROBU62404 ROBU62403 ROBU62402 ROBU62401 ROBU62400 ROBU62603 ROBU63502 ROBU63804 ROBU63803 ROBU63802 ROBU63801 ROBU63800 ROBU62803 ROBU63503 ROBU63501 ROBU63500 ROBU63004 ROBU63003 ROBU63002 ROBU63001 ROBU63000 ROBU62904 ROBU62903 ROBU62902 ROBU62901 ROBU63504 ROBU61504 ROBU61801 ROBU61800 ROBU61704 ROBU61703 ROBU61702 ROBU61701 ROBU61700 ROBU61604 ROBU61603 ROBU61602 ROBU61600 ROBU61804 ROBU61503 ROBU61502 ROBU61501 ROBU61500 ROBU61404 ROBU61403 ROBU61402 ROBU61401 ROBU61400 ROBU61304 ROBU61601 ROBU62100 ROBU62302 ROBU62301 ROBU62300 ROBU62204 ROBU62203 ROBU62202 ROBU62201 ROBU62200 ROBU62104 ROBU62103 ROBU61802 ROBU62101 ROBU16105 ROBU16115 ROBU16125 ROBU16205 ROBU16255 ROBU16305 ROBU16405 ROBU16505 ROBU16605 ROBU16705 ROBU16805 ROBU16905 ROBU16999 ROBU6130F ROBU6140F ROBU6150F ROBU6160F ROBU6170F ROBU6180F ROBU6190F ROBU6200F ROBU1610F ROBU1611F ROBU1612F ROBU1620F ROBU1625F ROBU1630F ROBU1640F ROBU1650F ROBU1660F ROBU1670F ROBU1680F ROBU1690F
VitraPOR® Glass Filter Discs, Biplane
Suodattimet Suodatinkiekot
VitraPOR® glass filters are precision-engineered porous glass components for effective filtration in laboratory and industrial settings.
  • Made from high-purity borosilicate glass 3.3
  • Available in different porosities
  • Chemically resistant and autoclavable
  • Reusable and environmentally friendly
  • Custom sizes and designs on request
  • Excellent flow rates and mechanical strength
ROBU biplane VitraPOR filters made of high-quality borosilicate glass are engineered for reliable and precise filtration across a wide range of laboratory and industrial applications.